Skip to Content
Merck
All Photos(2)

Documents

164603

Sigma-Aldrich

3-Nitropropionic acid

A cell-permeable, irreversible inactivator (~3 mM) of succinate dehydrogenase that upon oxidation by Complex II forms a covalent adduct with Arg297 in the active site of the enzyme.

Synonym(s):

3-Nitropropionic acid, 3-Nitropropionic acid, 3-NP, 3-NPA, 3NPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NO4
CAS Number:
Molecular Weight:
119.08
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

ethanol: 100 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C3H5NO4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)

InChI key

WBLZUCOIBUDNBV-UHFFFAOYSA-N

General description

A cell-permeable, irreversible inactivator (~3 mM) of succinate dehydrogenase that upon oxidation by Complex II forms a covalent adduct with Arg297 in the active site of the enzyme. Disrupts the blood-brain barrier and causes severe energy (ATP) impairment. Also replicates several pathophysiological symptoms of Huntington disease, including spontaneous choreiform and dystonic movements and degeneration of striatum in rodent models. Chronic administration of 3-NP (38 mg/kg/day) is shown to cause activation of c-Jun kinase in striatum.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Beal, M., et al. 1993, J. Neurosci.13, 4181.
Pang, Z., et al. 1997, J. Neuroscience17, 3064.
Huang, L., et al. 2006, J. Bio. Chem. 281, 5965.
Garcia, M., et al. 2002, J. Neurosci. 22, 2174.
Kim, G. W., et al. 2003, J. Neurosci.23, 8733.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service