Skip to Content
Merck
All Photos(1)

Documents

1.07913

Supelco

Boiling chips granules

size 2-8 mm

Synonym(s):

Boiling stones

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.21

size

2-8 mm

Quality Level

storage temp.

no temp limit

General description

~ 2-8 mm

Analysis Note

Substances soluble in water: ≤ 1

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service