Skip to Content
Merck
All Photos(1)

Documents

1.05812

Supelco

Magnesium foil

Synonym(s):

Magnesium foil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
MDL number:
UNSPSC Code:
12141502
EC Index Number:
231-104-6
NACRES:
NA.21

Assay

≥99.0% (complexometric)

Quality Level

form

solid

impurities

≤0.05% Substances insoluble in hydrochloric acid

bp

1107 °C/1013.25 hPa

mp

651 °C

density

1.75 g/cm3 at 20 °C

cation traces

Cu: ≤0.005%
Fe: ≤0.05%
Pb: ≤0.01%

storage temp.

no temp limit

InChI

1S/Mg/q+2

InChI key

JLVVSXFLKOJNIY-UHFFFAOYSA-N

Related Categories

Application

  • Moisture Battery Formed by Direct Contact of Magnesium with Foamed Polyaniline.: This study explores the formation of a moisture battery through the direct contact of magnesium foil with foamed polyaniline, showcasing its potential in energy storage applications (Xie et al., 2016).
  • High performance magnesium anode in paper-based microfluidic battery, powering on-chip fluorescence assay.: Research demonstrated the high performance of a magnesium anode within a paper-based microfluidic battery, emphasizing its application in powering on-chip fluorescence assays (Koo et al., 2014).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service