KEY267019955
Key Organics / BIONET
3H-spiro[1,3-benzothiazole-2,1'-cyclopentane]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
50 - 52
Iupac Name
3H-spiro[benzo[d]thiazole-2,1'-cyclopentane]
Inchi Code
1S/C11H13NS/c1-2-6-10-9(5-1)12-11(13-10)7-3-4-8-11/h1-2,5-6,12H,3-4,7-8H2
InChI key
VOYZBGVSNIQACE-UHFFFAOYSA-N
storage temp.
Store under inert gas
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/BE-0710
Melting Point
50 - 52
Signal Word
Warning
Hazard Codes
Precautionary statements
MSDS
https://www.keyorganics.net/sds/BE-0710
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.