Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249EC9D

J & W Pharmlab LLC

2-Bromo-5-ethyl-thiazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
192.08
MDL number:

Iupac Name

2-bromo-5-ethyl-1,3-thiazole

Inchi Code

1S/C5H6BrNS/c1-2-4-3-7-5(6)8-4/h3H,2H2,1H3

InChI key

GTUCNDIOXDOBPC-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Yellow Liquid

Product Link

https://www.jwpharmlab.com/home/product/90R0325

MSDS

https://www.jwpharmlab.com/home/product/90R0325


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library