Skip to Content
Merck
All Photos(1)

Key Documents

ENA465564472

Enamine

5H,6H,7H-imidazo[2,1-b][1,3]thiazin-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
156.21
MDL number:

mp

202-205

Iupac Name

6,7-dihydro-5H-imidazo[2,1-b][1,3]thiazin-6-ol

Inchi Code

1S/C6H8N2OS/c9-5-3-8-2-1-7-6(8)10-4-5/h1-2,5,9H,3-4H2

InChI key

NLCNMLXTZXABIW-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-246424

Melting Point

202-205

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-246424


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library