ENA341293200
Enamine
1-(trimethyl-1H-pyrazol-4-yl)propan-1-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-(1,3,5-trimethyl-1H-pyrazol-4-yl)-1-propanone
Inchi Code
1S/C9H14N2O/c1-5-8(12)9-6(2)10-11(4)7(9)3/h5H2,1-4H3
InChI key
XYCNZUWSRXFCSZ-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-88651
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-88651
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.