CO1H324AB51F
CombiPhos Catalysts Inc
4-Chloro-5-bromo-2-phenylpyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
269.53
Iupac Name
5-bromo-4-chloro-2-phenylpyrimidine
Inchi Code
1S/C10H6BrClN2/c11-8-6-13-10(14-9(8)12)7-4-2-1-3-5-7/h1-6H
InChI key
AFUIIMJXYMULOL-UHFFFAOYSA-N
Purity
95%
Country of Origin
US
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.