Skip to Content
Merck
All Photos(1)

Key Documents

CIAH98D1DF9A

ChemScene LLC (Preferred Partner)

3-(Pyridin-2-yl)propan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
136.2
MDL number:

bp

227.5+-23.0 C at 760 mmHg

Iupac Name

3-(2-pyridinyl)-1-propanamine

Inchi Code

1S/C8H12N2/c9-6-3-5-8-4-1-2-7-10-8/h1-2,4,7H,3,5-6,9H2

InChI key

UGYRJDSEKCYZKI-UHFFFAOYSA-N

storage temp.

4C, protect from light

Purity

98%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/15583-16-1.html

Boiling point

227.5+-23.0 C at 760 mmHg

Signal Word

Danger

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0006435.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library