Skip to Content
Merck
All Photos(1)

Key Documents

A-105

Supelco

N-Acetylretigabine solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16FN3O
CAS Number:
Molecular Weight:
273.31
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NC1=C(NC(C)=O)C=CC(NCC2=CC=C(F)C=C2)=C1

InChI

1S/C15H16FN3O/c1-10(20)19-15-7-6-13(8-14(15)17)18-9-11-2-4-12(16)5-3-11/h2-8,18H,9,17H2,1H3,(H,19,20)

InChI key

YGNSHYRMOVKKSL-UHFFFAOYSA-N

General description

A Certified Spiking Solution® suitable for use in retigabine testing methods by LC/MS or GC/MS for applications in clinical toxicology, medication monitoring, urine drug testing, pharmaceutical research, or forensic analysis. N-Acetylretigabine is the primary urinary metabolite of retigabine, a new antiepileptic drug developed for treatment of partial seizures. Also known as ezogabine, the drug is marketed under the trade names Potiga® in the US and Trobalt in the EU.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Potiga is a registered trademark of Valeant Pharmaceuticals North America LLC
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meijuan Zou et al.
Scientific reports, 6, 27331-27331 (2016-06-03)
Astrocyte elevated gene-1 (AEG-1) has been explored as a novel target for human glioma therapy, thus reflecting its potential contribution to gliomagenesis. In the present study, we investigated the effect of DYT-40, a novel synthetic 2-styryl-5-nitroimidazole derivative, on cell growth
Wei Zou et al.
Carbohydrate polymers, 152, 441-449 (2016-08-16)
The roles that the compact structure and proteins in pasta play in retarding evolution of starch molecular structure during in vitro digestion are explored, using four types of cooked samples: whole pasta, pasta powder, semolina (with proteins) and extracted starch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service