Skip to Content
Merck
All Photos(1)

Key Documents

ATC311765233

Atlantic Research Chemicals Ltd

2-Chloro-6-(trifluoromethyl)nicotinoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
244
MDL number:

Iupac Name

2-chloro-6-(trifluoromethyl)nicotinoyl chloride

Inchi Code

1S/C7H2Cl2F3NO/c8-5-3(6(9)14)1-2-4(13-5)7(10,11)12/h1-2H

InChI key

WPPJGHKCGNVACP-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=CC00223

MSDS

https://atlantic-chemicals.com/product/?code=CC00223


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library