Skip to Content
Merck
All Photos(1)

Key Documents

ATE517249803

AstaTech, Inc.

7-CHLORO-1-METHYL-1H-IMIDAZO[4,5-B]PYRIDINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
167.6
MDL number:

Iupac Name

7-chloro-1-methyl-1H-imidazo[4,5-b]pyridine

Inchi Code

1S/C7H6ClN3/c1-11-4-10-7-6(11)5(8)2-3-9-7/h2-4H,1H3

InChI key

MCOLKVLGCQWNBL-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=95284

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=91609


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library