ATE429354816
AstaTech, Inc.
(4-BROMOPHENYL)(THIOPHEN-2-YL)METHANONE
About This Item
Iupac Name
(4-bromophenyl)(2-thienyl)methanone
Inchi Code
1S/C11H7BrOS/c12-9-5-3-8(4-6-9)11(13)10-2-1-7-14-10/h1-7H
InChI key
DDUPBKRQEURUSF-UHFFFAOYSA-N
storage temp.
Purity
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=CS1413
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.