Skip to Content
Merck
All Photos(1)

Key Documents

APO458378685

Apollo Scientific Ltd

2-Chloro-6-iodobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
331.38
MDL number:

Iupac Name

2-(bromomethyl)-1-chloro-3-iodobenzene

Inchi Code

1S/C7H5BrClI/c8-4-5-6(9)2-1-3-7(5)10/h1-3H,4H2

InChI key

CDASKTNKXJMYDX-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Product Link

https://store.apolloscientific.co.uk/product/2-chloro-6-iodobenzyl-bromide

MSDS

https://store.apolloscientific.co.uk/product/2-chloro-6-iodobenzyl-bromide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library