Documents
Z693758
LabJaws all-position clamp holder
nickel plated
Synonym(s):
clamp holders, frame connectors
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
material
nickel plated
manufacturer/tradename
Troemner 916085
jaw capacity
0-19 mm
General description
Permits adjustment at any angle in any plane. Holders are set at 90° to each other, connected by a 90° connector, allowing 360° rotation.
Legal Information
LabJaws is a trademark of Henry Troemner LLC