Z530891
Flexible PTFE adapter
top female joint: ST/NS 24/40, bottom male joint: ST/NS 24/40
All Photos(3)
About This Item
Recommended Products
Joint
female joint: ST/NS top24/40
male joint: ST/NS bottom24/40
Related Categories
General description
Used for the transfer of powders and liquids. Can be compressed to approximately 190 mm or expanded to 250 mm. Glass joints with flexible corrugated PTFE tubing is pliable enough to create an offset adapter.
Other Notes
Picture is an example of male and female joint style.