Z159581
Cochranes Minit size bond tube sets
white, L 21.0 cm
Synonym(s):
chemistry model sets, molecular model sets
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
manufacturer/tradename
Cochranes 0288
L
21.0 cm
color
white
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service