Key Documents
Z131946
Nalgene® calcium chloride drying tube
About This Item
UNSPSC Code:
41100000
packaging
pkg of 12 ea
General description
PP, single bulb, tubulation at each end. Standard 8 in. length can be reduced to 6 in. or 4 in. by cutting with knife.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
comparable product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)