Documents
Z123323
Sigma-Aldrich
Paper insert
round hole, diam. 3.0 mm
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
material
round hole
technique(s)
IR spectroscopy: suitable
diam.
3.0 mm
General description
For making smaller KBr pellets. Use with the Aldrich macro-micro KBr pellet die (Z506699). Inserts also fit the Magnetic KBr pellet holder (Z112489) or Econo-Card (Z123544).