Skip to Content
Merck
All Photos(1)

Documents

W332700

Sigma-Aldrich

2-Acetyl-3,5(6)-dimethylpyrazine, mixture of isomers

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2O
CAS Number:
Molecular Weight:
150.18
FEMA Number:
3327
EC Number:
Council of Europe no.:
11294
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.055
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor pressure

3 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.517 (lit.)

bp

70 °C/7 mmHg (lit.)

density

1.073 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

hazelnut; nutty; roasted

SMILES string

CC(=O)c1ncc(C)nc1C

InChI

1S/C8H10N2O/c1-5-4-9-8(7(3)11)6(2)10-5/h4H,1-3H3

InChI key

UCGOSAWBWFUKDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service