W270105
4-(3,4-Methylenedioxyphenyl)-2-butanone
≥99%
Synonym(s):
Piperonyl acetone
About This Item
Recommended Products
biological source
synthetic
Assay
≥99%
bp
176 °C/17 mmHg (lit.)
mp
49-54 °C (lit.)
application(s)
flavors and fragrances
SMILES string
CC(=O)CCc1ccc2OCOc2c1
InChI
1S/C11H12O3/c1-8(12)2-3-9-4-5-10-11(6-9)14-7-13-10/h4-6H,2-3,7H2,1H3
InChI key
TZJLGGWGVLADDN-UHFFFAOYSA-N
Other Notes
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service