Skip to Content
Merck
All Photos(1)

Key Documents

W256110

Sigma-Aldrich

trans-2-Hexen-1-al

natural, ≥95%, FG

Synonym(s):

trans-2-Hexenal, Leaf aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCHO
CAS Number:
Molecular Weight:
98.14
FEMA Number:
2560
Beilstein:
1699684
EC Number:
Council of Europe no.:
748
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.073

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

3.4 (vs air)

vapor pressure

10 mmHg ( 20 °C)

Assay

≥95%

refractive index

n20/D 1.446 (lit.)

bp

47 °C/17 mmHg (lit.)

density

0.846 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

almond; apple; green; minty; vegetable

storage temp.

2-8°C

SMILES string

CCC\C=C\C=O

InChI

1S/C6H10O/c1-2-3-4-5-6-7/h4-6H,2-3H2,1H3/b5-4+

InChI key

MBDOYVRWFFCFHM-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.9 °F - closed cup

Flash Point(C)

38.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Jung Joo et al.
International journal of food microbiology, 153(3), 297-305 (2011-12-20)
Trans-2-hexenal, a naturally occurring plant volatile with antimicrobial capacity, was encapsulated into β-cyclodextrins (β-CDs), enzymatically modified starch, and shown effective to control main microorganisms causing food spoilage (Alternaria solani, Aspergillus niger, Botrytis cinerea, Colletotrichum acutatum, Penicillium sp). Loaded β-CDs were
Jaroslav Flesar et al.
Veterinary microbiology, 145(1-2), 129-133 (2010-04-23)
In total, 26 natural compounds of various chemical classes (flavonoids, alkaloids, terpenoids) and 19 crude extracts from selected plants were tested in vitro for antibacterial activity against three strains of P. larvae, the causal agent of American Foulbrood Disease of
Reiko Kiwamoto et al.
Chemical research in toxicology, 25(12), 2630-2641 (2012-09-18)
trans-2-Hexenal (2-hexenal) is an α,β-unsaturated aldehyde that occurs naturally in a wide range of fruits, vegetables, and spices. 2-Hexenal as well as other α,β-unsaturated aldehydes that are natural food constituents or flavoring agents may raise a concern for genotoxicity due
G S Germinara et al.
Journal of food protection, 75(2), 366-370 (2012-02-01)
The purpose of this work was to develop a biodegradable carrier material to control insect pests in cereal products. To this aim, (E)-2-hexenal was used, being a natural compound with antimicrobial activity that is also commonly adopted as a flavoring
Kensuke Sakamaki et al.
Journal of agricultural and food chemistry, 59(12), 6667-6671 (2011-05-10)
A test apple beverage made up of apple juice (20%), high-fructose corn syrup (11.5%), citric acid (0.43%), trisodium citrate (0.02%), apple-odor flavor (0.1%), and ascorbic acid (0.02%) was stored at 40 °C and then analyzed for the change of odor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service