Documents
SYNP130100M
Synthware peptide synthesis vessel with fritted disc and T-bore PTFE stopcock
100 mL, porosity: medium, GL 25
Synonym(s):
Synthware P130100M, Solid phase peptide synthesis vessel
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P130100M
capacity
100 mL
100 mL
fitting
GL 25
Related Categories
General description
With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Legal Information
Synthware is a trademark of Kemtech America Inc.