Documents
SYNF806150
Synthware Buchner funnel with perforated plate
150 mL, plate diam. 60 mm
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F806150
capacity
150 mL
150 mL
plate diam.
60 mm
General description
With sealed-in perforated glass disc to support filter media. Pressure may be used but should not exceed 15 psi. This design solve the contamination problem with fritted disc and also the blindness problem with ceramic Buchner funnel.
Legal Information
Synthware is a trademark of Kemtech America Inc.