SYNF164100
Synthware™ two neck reaction flask with glass stopcock
capacity 100 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F164100, Glass reaction flask
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass flask
round bottom
two-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F164100
capacity
100 mL
Related Categories
General description
With a 2 mm glass stopcock having a constant pressure retention device.
Legal Information
Synthware is a trademark of Kemtech America Inc.