N2875
Naphthol AS-D acetate
Synonym(s):
3-(Acetyloxy)-N-(2-methylphenyl)-2-naphthalenecarboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H17NO3
CAS Number:
Molecular Weight:
319.35
Beilstein:
2666666
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CC(=O)Oc1cc2ccccc2cc1C(=O)Nc3ccccc3C
InChI
1S/C20H17NO3/c1-13-7-3-6-10-18(13)21-20(23)17-11-15-8-4-5-9-16(15)12-19(17)24-14(2)22/h3-12H,1-2H3,(H,21,23)
InChI key
PMRFWCJXRSRVPS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Substrates
Substrate for histochemical localization of esterases
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service