N21006
3-Nitrophenyl disulfide
98%
Synonym(s):
Bis(3-nitrophenyl) disulfide, Nitrophenide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(O2NC6H4)2S2
CAS Number:
Molecular Weight:
308.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
98%
mp
78-80 °C (lit.)
SMILES string
[O-][N+](=O)c1cccc(SSc2cccc(c2)[N+]([O-])=O)c1
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J J Allocco et al.
The Journal of parasitology, 87(6), 1441-1448 (2002-01-10)
Unsporulated oocysts of the protozoan parasite Eimeria tenella contain high levels of mannitol, which is thought to be the principal energy source for the process of sporulation. Biosynthesis and utilization of this sugar alcohol occurs via a metabolic pathway known
D M Schmatz
Parasitology, 114 Suppl, S81-S89 (1997-01-01)
A metabolic pathway known as the mannitol cycle has been identified in Eimerian parasites. The pathway is a shunt off of the glycolytic pathway at fructose-6-phosphate (F6P). Two enzymes convert F6P to mannitol and two other enzymes are responsible for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service