Skip to Content
Merck
All Photos(1)

Documents

GF28998938

Brass alloy, Cu63Zn37

200mm tube, 1.18mm outside diameter, 0.23mm wall thickness, 0.72mm inside diameter, as drawn

Synonym(s):

CU027170

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
30102010
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 289-989-38

L × wall thickness

200 mm × 0.23 mm

O.D. × I.D.

1.18 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service