Skip to Content
Merck
All Photos(1)

Documents

DWK63B93P

Kimble® Disposable Pasteur Pipet

L 9.0 in., soda-lime glass, Cotton-plugged

Synonym(s):

63B93P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.04

material

soda-lime glass

feature

Cotton-plugged

packaging

pkg of 1000 ea

manufacturer/tradename

Kimble® 63B93P

L

9.0 in.

Looking for similar products? Visit Product Comparison Guide

Application

Bacteriology, immunology, hematology, and serology

Features and Benefits

Removes risk of cross-contamination of high value specimens and laboratory reagents.

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service