CLS9944512
Pyrex® disposable culture tubes, rimless
capacity 6.0 mL
Synonym(s):
culture tube, glass tubes, test tubes, Corning® 99445
About This Item
Recommended Products
material
borosilicate glass
round bottom tube
description
dispenser arrangement
sterility
non-sterile
feature
cap: no
packaging
case of 1000
pkg of 1000 ea
pack of 250 ea
manufacturer/tradename
Corning 99445-12
O.D. × L
12 mm × 75 mm
capacity
6.0 mL
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service