Skip to Content
Merck
All Photos(1)

Documents

BR43070

BRAND® glass bottle-top tilt pipette dispenser

without bottle

Synonym(s):

Automatic pipettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

Calibrated to deliver (TD, Ex)
without bottle

manufacturer/tradename

BRAND 43070

volume

25 mL

color

green markings

fitting

(29/32)

Looking for similar products? Visit Product Comparison Guide

General description

Calibrated to deliver (TD, Ex). Inscriptions in green enamel. The automatic dispenser is particularly well-suited for dispensing suspensions. Delivery range approximately +/- 10%.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service