BR130874
BRAND® storage bottle
volume 10 L, narrow neck
Synonym(s):
Bottles, Storage Bottles
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
high-density polyethylene
narrow-mouth bottle
round bottle
white bottle
Joint
joint threaded neck
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 130874
capacity
10 L
diam. × H
210 mm × 420 mm
volume
10 L
10000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With screw cap and carrying handle.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service