BAF131930500
Poxygrid® benchtop bag holder kit
includes 1 bag holder, 1 cover & 100 bags
Synonym(s):
bag holder
All Photos(3)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F13193-0500
Related Categories
General description
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
Features and Benefits
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
- Rectangular steel wire frame holds a polyethylene bag 22 x 28cm (8¹/₂ x 11"); coated in easy to clean, durable, orange color epoxy
- Snug fitting polypropylene plastic cover helps hold the bag securely in place
- Reusable; can be cleaned by hand or in automated dish washers; autoclavable
- Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes
- Kit includes 100 Bench Top Biohazard Bags (F13166-0000); can also be used with Clavies Autoclavable Bags (H13182-0812)
Legal Information
Poxygrid is a registered trademark of SP Scienceware, a division of SP