AEM2A2022
Xion AEM-Pention-72-5CL
thickness 20 μm, L × W 5 cm × 5 cm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
solid
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
L × W
5 cm × 5 cm
thickness
20 μm
greener alternative category
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for use as Proton-Exchange Membranes (Fuel-Cell Membranes). Click here for more information.
Application
XION Composite PENTION Membranes are ultra-thin, ultra-strong, and provide ultra-high performance while demonstrating highly stable performance as reinforced anion exchange membranes (AEM). The ionomer structure contains a poly(norbornene) backbone with quaternary ammonium functional groups. A reinforcement layer is integrated into the structure of the membrane to provide enhanced mechanical properties. The enhanced mechanical properties as free-standing membranes, providing higher ionic conductance without sacrificing strength. Applications for XION Composite PENTION Membranes include fuel cells, electrolysis, and flow batteries.
Legal Information
Product of Xergy
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service