Skip to Content
Merck
All Photos(3)

Key Documents

937541

Sigma-Aldrich

Ethyl 3-amino-3-(pyridin-2-yl)acrylate

≥95%

Synonym(s):

3-Amino-3-(2-pyridinyl)-2-propenoic acid ethyl ester, Ethyl 3-amino-3-(2-pyridinyl)-2-propenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O2
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352142
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder or crystals

color

white to yellow, Beige

functional group

acrylate
amine
ester

storage temp.

2-8°C

SMILES string

O=C(OCC)C=C(N)C1=NC=CC=C1

InChI

1S/C10H12N2O2/c1-2-14-10(13)7-8(11)9-5-3-4-6-12-9/h3-7H,2,11H2,1H3

InChI key

YYJBOEYBEJCPES-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service