Skip to Content
Merck
All Photos(1)

Documents

931667

Sigma-Aldrich

Fmoc-L-F4G(Pbf,Sub)-OH

Novabiochem®, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H52N4O7S
Molecular Weight:
889.07
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

composition

May contain ≤10% chloroform

storage temp.

−20°C

Application

Fmoc-L-F4G(Pbf,Sub)-OH is an arginine analog that can be used to prepare novel peptides for usage as putative therapeutics or in the elucidation of structure-activity relationships.

Legal Information

Product of Peptidream
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service