Skip to Content
Merck
All Photos(3)

Documents

915165

Sigma-Aldrich

[Pd(IPr)(3-CF3-AN)Cl2]

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H42Cl2F3N3Pd
Molecular Weight:
727.04
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

[Pd(IPr)(3-CF3-AN)Cl2] is a Pd-catalyst that can be used in a broad range of reactions, including cross-coupling, C-H functionalization, electrophilic additions. NHC-Pd-aniline complexes of this type have many advantages, including higher reactivity in cross-coupling reactions than similar complexes using cinnamyl and pyridine throw-away ligands and simple reaction tunability through substituion of aniline ligand.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service