Skip to Content
Merck
All Photos(1)

Documents

901130

Sigma-Aldrich

1,2-Bis(2-azidoethoxy)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N6O2
CAS Number:
Molecular Weight:
200.20
MDL number:
UNSPSC Code:
12352202

form

liquid

Quality Level

refractive index

n20/D 1.47

density

1.15 g/mL

storage temp.

2-8°C

SMILES string

[N-]=[N+]=NCCOCCOCCN=[N+]=[N-]

InChI

1S/C6H12N6O2/c7-11-9-1-3-13-5-6-14-4-2-10-12-8/h1-6H2

InChI key

OHZGAFKSAANFAS-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service