Skip to Content
Merck
All Photos(1)

Key Documents

900557

Sigma-Aldrich

Rare earth doped phosphor nanoparticles

fluorescence λem 620 nm (main peak), 50 mg/mL in H2O

Synonym(s):

CANdot® Series X red, Fluorescent nanoparticles, Luminescent nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303

form

dispersion
nanoparticles

composition

Y(P,V)O4:Eu

concentration

50 mg/mL in H2O

particle size

10 nm

fluorescence

λem 620 nm (main peak)
λem 620 nm±15 nm

functional group

ethylene glycol

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Cd-free phosphor nanoparticles find applications in solid state lighting, sensors, security pigments etc. Our downshifting rare earth doped fluorescent nanoparticles are based on inorganic host lattices like sulfates, phosphates and vanadates which can be excited by a UV light source with strong emission at 275 nm. The emission characteristics of these nanoparticles have been tuned with dopants such as Bismuth, Cerium, Europium, Terbium etc. and are useful for the aforementioned applications.

Legal Information

Fraunhofer CAN is a research division of the Fraunhofer IAP
CANdot is a registered trademark of Fraunhofer CAN

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Color tuning of indium phosphide quantum dots for cadmium-free quantum dot light-emitting devices with high efficiency and color saturation.
Ippen C, et al.
Society for Information Display International Symposium Digest of Technical Papers, 23, 285-293 (2015)
Optical behaviour of cadmium and mercury free eco-friendly lamp nanophosphor for display devices.
Tiwari R, et al.
Results in Physics, 4, 63-68 (2014)
Influence of Structural Polymorphs on Tunable White Light Generation from Orange-Red-Emitting BiPO4:Eu3+ Phosphor by Surface Modification.
Muller JG, et al.
The Journal of Physical Chemistry C, 118, 19308-19314 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service