Skip to Content
Merck
All Photos(1)

Key Documents

860425

Sigma-Aldrich

Myristyltrimethylammonium bromide

99%

Synonym(s):

Tetradecyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Br)(CH3)3
CAS Number:
Molecular Weight:
336.39
Beilstein:
3633227
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

mol wt

micellar avg mol wt 27,000

aggregation number

80

CMC

4-5 mM (20-25°C)

mp

245-250 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1

InChI key

CXRFDZFCGOPDTD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cationic detergent, antiseptic.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihaela Necula et al.
The Journal of biological chemistry, 282(14), 10311-10324 (2007-02-08)
Alzheimer disease is characterized by the abnormal aggregation of amyloid beta peptide into extracellular fibrillar deposits known as amyloid plaques. Soluble oligomers have been observed at early time points preceding fibril formation, and these oligomers have been implicated as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service