Skip to Content
Merck
All Photos(2)

Key Documents

808873

Sigma-Aldrich

Pyromellitic dianhydrate dimethacrylate

(1:1 meta:para isomer mixture), contains 4-methoxyphenol as inhibitor

Synonym(s):

PMDM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22O12
Molecular Weight:
478.40
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

form

powder

contains

4-methoxyphenol as inhibitor

mp

118 °C

SMILES string

OC(C1=CC(C(OCCOC(C(C)=C)=O)=O)=C(C(O)=O)C=C1C(OCCOC(C(C)=C)=O)=O)=O.OC(C2=CC(C(O)=O)=C(C(OCCOC(C(C)=C)=O)=O)C=C2C(OCCOC(C(C)=C)=O)=O)=O

InChI

1S/2C22H22O12/c1-11(2)19(27)31-5-7-33-21(29)15-9-14(18(25)26)16(10-13(15)17(23)24)22(30)34-8-6-32-20(28)12(3)4;1-11(2)19(27)31-5-7-33-21(29)15-10-16(14(18(25)26)9-13(15)17(23)24)22(30)34-8-6-32-20(28)12(3)4/h2*9-10H,1,3,5-8H2,2,4H3,(H,23,24)(H,25,26)

InChI key

KOLVKKCMPOWKSN-UHFFFAOYSA-N

General description

1:1 mixture of meta/para isomers.

Application

May be used as a dental adhesive functional monomer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combinatorial investigation of structure-properties relationships and microcosmic curing mechanism of dental adhesives functional monomers.
Sun, et al.
Vibrational Spectroscopy, 51, 93-99 (2009)
Francisco López-Suevos et al.
Dental materials : official publication of the Academy of Dental Materials, 24(6), 778-785 (2007-11-06)
Evaluate the effects of core structure and storage conditions on the mechanical properties of acid-resin modified composites and a control material by three point bending and conversion measurements 15min and 24h after curing. The monomers pyromellitic dimethacrylate (PMDM), biphenyldicarboxylic-acid dimethacrylate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service