Skip to Content
Merck
All Photos(1)

Key Documents

793884

Lithium bis(fluorosulfonyl)imide

ultra dry, 99.9% trace metals basis

Synonym(s):

LiFSI, Lithium bis(fluorosulfonyl)amide, Lithium bis(fluorosulfonyl)imido, Lithium imidodisulfuryl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
F2LiNO4S2
CAS Number:
Molecular Weight:
187.07
UNSPSC Code:
41116105
PubChem Substance ID:

Assay

99.9% trace metals basis

form

solid

SMILES string

FS([N-]S(F)(=O)=O)(=O)=O.[Li+]

InChI

1S/F2NO4S2.Li/c1-8(4,5)3-9(2,6)7;/q-1;+1

InChI key

VDVLPSWVDYJFRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

LiFSI is a promising new electrolyte additive for lithium ion batteries showing increased conductivity over LiPF6. Bis(fluorosulfonyl)imide has also been tested in next generation gel polymer electrolytes with improved safety properties

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guerfi; A.;
Journal of Solid State Electrochemistry, 13(7), 1003-1014 (2009)
Han; Hong-Bo;
Journal of Power Sources, 196(7), 3623-3632 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service