Skip to Content
Merck
All Photos(1)

Documents

791458

Sigma-Aldrich

EL-HTE high temperature electrolyte

Synonym(s):

EL-HTE, EL-HTE electrolyte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid

conductivity

2-4 mS/cm at 20 °C

Application

  • EL-HTE - High Temperature Electrolyte is a liquid electrolyte to achieve ultra-long life in DSC systems while minimising solvent evaporation.
  • EL-HTE High Temperature Electrolyte has a lower vapour pressure than EL-HSE High Stability Electrolyte but is otherwise chemically very similar.
  • EL-HTE High Temperature Electrolyte is particularly suitable for flexible DSC;where transparent flexible substrates do not offer the same hermetic barrier properties such as glass or metal substrates.

Legal Information

Dyesol® Product
Dyesol is a registered trademark of Greatcell Solar

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.0 °F

Flash Point(C)

87.78 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service