Skip to Content
Merck
All Photos(1)

Key Documents

779652

Sigma-Aldrich

(R)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthalene

≥95.0% (HPLC)

Synonym(s):

(R)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16Br2O2
CAS Number:
Molecular Weight:
472.17
Beilstein:
4712472
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥95.0% (HPLC)

form

lumps

optical purity

enantiomeric excess: ≥98.0%

SMILES string

COc1c(Br)cc2ccccc2c1-c3c(OC)c(Br)cc4ccccc34

InChI

1S/C22H16Br2O2/c1-25-21-17(23)11-13-7-3-5-9-15(13)19(21)20-16-10-6-4-8-14(16)12-18(24)22(20)26-2/h3-12H,1-2H3

InChI key

DFTUKDIMHCCQIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service