Skip to Content
Merck
All Photos(1)

Key Documents

779466

Sigma-Aldrich

3-(2,4,6-Trimethylphenyl)-5,6,7,8-tetrahydro-4H-cycloheptathiazol-3-ium perchlorate

≥95% (HPLC)

Synonym(s):

5,6,7,8-Tetrahydro-3-(2,4,6-trimethylphenyl)-4H-cycloheptathiazolium perchlorate, Isa-Carbene, Isa-NHC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22ClNO4S
CAS Number:
Molecular Weight:
371.88
Beilstein:
19104993
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥95% (HPLC)

impurities

8.19-9.05% sulfur content

SMILES string

[O-]Cl(=O)(=O)=O.Cc1cc(C)c(c(C)c1)-[n+]2csc3CCCCCc23

InChI

1S/C17H22NS.ClHO4/c1-12-9-13(2)17(14(3)10-12)18-11-19-16-8-6-4-5-7-15(16)18;2-1(3,4)5/h9-11H,4-8H2,1-3H3;(H,2,3,4,5)/q+1;/p-1

InChI key

NYWYDMVZQBQLOV-UHFFFAOYSA-M

Application

Catalyst

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiichi Hirano et al.
Journal of the American Chemical Society, 131(40), 14190-14191 (2009-10-08)
An intramolecular N-heterocyclic carbene (NHC)-catalyzed hydroacylation of unactivated double bonds is reported. Systematic variation of the catalyst structure revealed an N-mesitylthiazolylidene annulated with a seven-membered ring to be especially reactive. This NHC enables a unique C-C bond-forming reaction to afford
Akkattu T Biju et al.
Journal of the American Chemical Society, 132(17), 5970-5971 (2010-04-14)
The N-heterocyclic carbene (NHC)-catalyzed hydroacylation of unactivated alkynes to provide alpha,beta-unsaturated ketones is reported. In addition, a rare case of an efficient and selective dually NHC-catalyzed cascade reaction involving the hydroacylation of alkynes and a subsequent intermolecular Stetter reaction allows
Raphaël Lebeuf et al.
Organic letters, 10(19), 4243-4246 (2008-09-04)
A large range of benzoins was successfully applied as C-nucleophiles in the palladium-catalyzed allylic alkylation with several allyl acetates, resulting in functionalized tertiary homoallylic alcohols. A number of unsymmetrical benzoins can be coupled with high levels of regio- and chemoselectivity.
Diastereoselective Synthesis of Trifluoromethylated γ-Butyrolactones via N-Heterocyclic Carbene-catalyzed Conjugated Umpolung of a,?-unsaturated Aldehydes.
Hirano, K., et al.
Advanced Synthesis & Catalysis, 350, 984-988 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service