Skip to Content
Merck
All Photos(1)

Key Documents

774596

Sigma-Aldrich

Bismuth oxide - yttria stabilized

contains 25 mol % yttria as stabilizer, 99.99% trace metals basis

Synonym(s):

25YSB, Bismuth yttrium oxide, Yttrium bismuth oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Bi2O3)0.75(Y2O3)0.25
CAS Number:
UNSPSC Code:
12352200

Assay

99.99% trace metals basis

form

powder

contains

25 mol % yttria as stabilizer

Application

Mixed ionic-electronic conductors based on bismuth oxide having a fluorite structure are attractive materials for solid-state ionic devices and electrochemical processes at intermediate to low temperatures. 25 mol% Y2O3 stabilized Bi2O3 (25YSB) founds applications as electrolyte material for intermediate temperature solid oxide fuel cell (IT-SOFC). Composites incorporating yttria-stabilized bismuth oxide (YSB) have also been investigated as cathode material for SOFC; owing to the high oxygen ionic conductivity and catalytic activity of YSB.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xia; C.;
Applied Physics Letters, 82(6), 901-901 (2003)
Kruidhof; H.;
Solid State Ionics, 37(2-3), 213-213 (1990)
B García-Baños et al.
Scientific reports, 9(1), 10809-10809 (2019-07-28)
In this study, real-time and in-situ permittivity measurements under intense microwave electromagnetic fields are proposed as a powerful technique for the study of microwave-enhanced thermal processes in materials. In order to draw reliable conclusions about the temperatures at which transformations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service