77301
(+)-Perillaaldehyde
≥98.0% (sum of enantiomers, GC)
Synonym(s):
(R)-4-Isopropenylcyclohexene-1-carboxaldehyde, (R)-p-Mentha-1,8-dien-7-al
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2413108
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Assay
≥98.0% (sum of enantiomers, GC)
optical activity
[α]20/D +136.0±2°, c = 10% in ethanol
optical purity
enantiomeric ratio: ≥99:1 (GC)
refractive index
n20/D 1.505
density
0.966 g/mL at 20 °C (lit.)
SMILES string
CC(=C)[C@@H]1CCC(C=O)=CC1
Other Notes
Highly purified (+)-perillaaldehyde useful as chiral building block
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T. Kitahara et al.
Tetrahedron, 44, 4713-4713 (1988)
C. Siegel et al.
The Journal of Organic Chemistry, 56, 6865-6865 (1991)
F. Kido et al.
Journal of the Chemical Society. Perkin Transactions 1, 229-229 (1992)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service