Skip to Content
Merck
All Photos(1)

Key Documents

763985

Sigma-Aldrich

2,2′-(9,9-Dioctyl-9H-fluorene-2,7-diyl)bisthiophene

97%

Synonym(s):

2,7-Bis(thien-2-yl)-9,9-dioctylfluorene, Thiophene, 2,2′-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H46S2
CAS Number:
Molecular Weight:
554.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

35-40 °C

SMILES string

CCCCCCCCC1(CCCCCCCC)c2cc(ccc2-c3ccc(cc13)-c4cccs4)-c5cccs5

InChI

1S/C37H46S2/c1-3-5-7-9-11-13-23-37(24-14-12-10-8-6-4-2)33-27-29(35-17-15-25-38-35)19-21-31(33)32-22-20-30(28-34(32)37)36-18-16-26-39-36/h15-22,25-28H,3-14,23-24H2,1-2H3

InChI key

HGAWLLXSUXWDFN-UHFFFAOYSA-N

Application

Fluorene monomer which contains thiophene designed to expand the
spectral width of absorption appropriate for sunlight harvesting in polymer solar cells.1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Functional Materials, 18, 2824-2833 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service