Skip to Content
Merck
All Photos(1)

Documents

759139

Sigma-Aldrich

β-N-Methyl-d3-amino-DL-alanine-15N2

98 atom % 15N, 98 atom % D, 97% (CP)

Synonym(s):

BMAA-15N2,d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(D)315NHCH2CH(15NH2)COOH
Molecular Weight:
123.14
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % 15N
98 atom % D

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

SMILES string

OC(C([15NH2])C[15N]([H])C([2H])([2H])[2H])=O

InChI

1S/C4H10N2O2/c1-6-2-3(5)4(7)8/h3,6H,2,5H2,1H3,(H,7,8)/i1D3,5+1,6+1

InChI key

UJVHVMNGOZXSOZ-OISARLQDSA-N

Related Categories

Packaging

This product is a Neurotoxin
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Caution

Neurotoxin

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service