Skip to Content
Merck
All Photos(1)

Documents

758108

Sigma-Aldrich

6-Methoxy-1,3-benzodioxole-5-carbaldehyde

97%

Synonym(s):

5-Formyl-6-methoxy-1,3-benzodioxole, 6-Methoxypiperonal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O4
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

109-115 °C

SMILES string

COc1cc2OCOc2cc1C=O

InChI

1S/C9H8O4/c1-11-7-3-9-8(12-5-13-9)2-6(7)4-10/h2-4H,5H2,1H3

InChI key

NCIYDDIPUGCVKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service